주요 문서
Z126454
Automatic shut-off quick-disconnect
coupling bodies, tube to pipe thread, bore 1/4 in.
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.35
제조업체/상표
CRC Press
보어(bore)
1/4 in.
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Spring-loaded, locking, made of PP with 316 SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically, making couplings excellent for handling air-sensitive liquids. Available for flexible tubing or rigid PE and nylon tubing using a ferruleless nut. Max. pressure: 90 psi at 70°F. Working temp.: 32 to 230°F (air/water). Use same bore size for coupling insert and body.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)