주요 문서
W318108
Sigma-Aldrich
o-Methoxycinnamaldehyde
≥96%, FG
로그인조직 및 계약 가격 보기
About This Item
Linear Formula:
CH3OC6H4CH=CHCHO
CAS Number:
Molecular Weight:
162.19
FEMA Number:
3181
EC Number:
유럽평의회 번호:
571
MDL number:
UNSPSC 코드:
12164502
PubChem Substance ID:
플래비스(Flavis) 번호:
5.048
NACRES:
NA.21
생물학적 소스
synthetic
Quality Level
Grade
FG
Halal
Kosher
규정 준수
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515
분석
≥96%
bp
160-161 °C/12 mmHg (lit.)
mp
44.0-49.0 °C (lit.)
응용 분야
flavors and fragrances
문건
see Safety & Documentation for available documents
식품 알레르기항원
no known allergens
감각 수용성의
cinnamon; woody; spicy; sweet
SMILES string
[H]C(=O)C=Cc1ccccc1OC
InChI
1S/C10H10O2/c1-12-10-7-3-2-5-9(10)6-4-8-11/h2-8H,1H3/b6-4+
InChI key
KKVZAVRSVHUSPL-GQCTYLIASA-N
유사한 제품을 찾으십니까? 방문 제품 비교 안내
픽토그램
Exclamation mark
신호어
Warning
유해 및 위험 성명서
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
표적 기관
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
235.4 °F
Flash Point (°C)
113 °C
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
이미 열람한 고객
Slide 1 of 1
1 of 1
Daishi Yamakawa et al.
Biochemical and biophysical research communications, 415(1), 174-180 (2011-10-29)
Blood vessels are mainly composed of intraluminal endothelial cells (ECs) and mural cells adhering to the ECs on their basal side. Immature blood vessels lacking mural cells are leaky; thus, the process of mural cell adhesion to ECs is indispensable
Ludger Wennemann et al.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen), 67(3), 499-509 (2003-04-17)
High wheel tractor applications of 4-methoxycinnamaldehyde (MCA)-coated corn granules ('grits') were conducted in Ruski Krstur (Serbia) in summer 2001 in a 5 ha corn field. Grits are a by-product after corn is harvested and separated from the cob and used
Jian-You Guo et al.
Biological & pharmaceutical bulletin, 29(11), 2214-2221 (2006-11-02)
Prostaglandin E2 (PGE2) works as a common final mediator of the febrile. Guizhi-Tang, one of the most famous traditional Chinese medical formula used to treat influenza, common cold and other pyretic conditions, was previously reported to reduce the production of
Jian-you Guo et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(13), 1087-1090 (2006-10-20)
To observe the effect of 2-methoxycinnamaldehyde (isolated from fraction A of Guizhi Tang) on activity of COX and PGE2 release in rat cerebral microvascular endothelial cells (rCMEC) stimulated by IL-1. rCMEC were cultured, and identified by immunohistochemistry for von Willebrand
R Giordani et al.
Phytotherapy research : PTR, 20(1), 58-61 (2006-01-07)
The antifungal activity of the essential oil from Cinnamomum cassia, alone or combined with amphotericin B, a drug widely used for most indications despite side-effects was investigated. The composition of the oil was analysed by GC/MS and characterized by its