TEMFF150CU
TEM-FF150CU
동의어(들):
FORMVAR FILM MESH CU 25/BX
1 of 10
Sigma-Aldrich
930318
Formvar Supported Copper Square Mesh
930245
Formvar/Carbon Supported Copper Grids
Millipore
RAWG0250C
MCE Membrane Filter, 1.2 μm Pore Size
TEM-FFT200CU25
Formvar Supported Copper Thin Bar Square Mesh
930261
SAMP2HNNB
Millex™ filter for the Samplicity® G2 System
AP408X105
Glass Fiber Membrane Filter, 0.7 μm Pore Size
SAMP2GNNB
RAWG04700
AQFA8X105
Quartz Membrane Filter
box of 25
150 mesh
유사한 제품을 찾으십니까? 방문 제품 비교 안내
Electronic Materials
Substrates & Electronic Components
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.