주요 문서
S679070
O-TERT-BUTYL-L-HYDROXYPROLINE TERT-BUTYL ESTER DIBENZENESULFONAMIDE SALT
AldrichCPR
로그인조직 및 계약 가격 보기
크기 선택
S679070-250MG
₩201,500
₩201,500
예상 입고일2025년 3월 07일세부사항
크기 선택
보기 변경
S679070-250MG
₩201,500
About This Item
Linear Formula:
C25H36N2O7S2
CAS Number:
Molecular Weight:
540.703
MDL number:
₩201,500
예상 입고일2025년 3월 07일세부사항
기타 정보
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)