DWK116200300
DWK11-6200-300
DWK11-6200-300-1PK
₩221,473
1 of 10
DWK11-6200-101
Wheaton® Microliter Vials With Inserts Installed
Supelco
1.14901
Flat-bottomed long tubes with screw caps
DWK11-5200-101
29391-U
Certified QSertVial™ (vial with fused-in insert) kit, screw thread, 12 x 32 mm, 0.3 mL, pkg of 100
CLS9944816
Pyrex® disposable culture tubes, screw-cap style, flat bottom
DWK73500-650
KIMBLE® disposable rimless culture tube
1.14902
Flat-bottomed tubes with screw caps
CLS8422100
Pyrex® heavy-duty round-bottom centrifuge tubes, with phenolic screw cap and rubber liner
29378-U
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
29307-U
Certified CD™ Vial (Center Draining) Kit, 9 mm thread with cap/septa, unassembled
amber vial
vial type with Marking Patch with 11-0000-300 inserts
pkg of 1 packs
WHEATON® 11-6200-300
유사한 제품을 찾으십니까? 방문 제품 비교 안내
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.