DWK116200101
DWK11-6200-101
DWK11-6200-101-1PK
₩329,063
1 of 10
Supelco
SU860097
Headspace vial, screw top, rounded bottom (vial only)
Sigma-Aldrich
Z250201
Wheaton wide-mouth bottle
P9029
o-Phenylenediamine
Z740940
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
1.09535
pH-indicator strips pH 0 - 14 Universal indicator
29378-U
Certified Kits, screw thread vials, 12 x 32 mm, 9 mm thread, unassembled, pkg of 100
854190
Vials, screw top, clear glass (vial only)
1.09435
Buffer solution
Z256293
Wheaton sample vials with rubber lined caps
854181-U
Headspace vials, flat top, rounded bottom, pkg of 100
amber vial
vial type with Marking Patch with 11-0000-101 inserts
pkg of 1 packs
WHEATON® 11-6200-101
유사한 제품을 찾으십니까? 방문 제품 비교 안내
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.