주요 문서
CAD4039
Standard point hypodermic needles, 304 stainless steel
gauge 23, L 1 1/2 in., nickel plated hub
로그인조직 및 계약 가격 보기
크기 선택
12 EA
₩191,125
₩191,125
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
12 EA
₩191,125
About This Item
UNSPSC 코드:
42142523
NACRES:
NB.03
₩191,125
구입 가능 여부는 고객센터에 문의하십시오.
material
304 stainless steel needle
nickel plated hub
포장
pack of 12 ea
제조업체/상표
Cadence Science 4039
길이
1 1/2 in.
게이지
23
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)