주요 문서
BAH369401010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 1.25 mL , ¼tsp, L × W 17.8 cm (7 in.) × 0.8125 in. (20 mm), sterile, pack of 10 ea (double bagged)
동의어(들):
Lab Scoop, dispensing tool, spoon
로그인조직 및 계약 가격 보기
크기 선택
10 EACH
₩146,566
₩146,566
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
10 EACH
₩146,566
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.35
₩146,566
구입 가능 여부는 고객센터에 문의하십시오.
material
white polystyrene
무균
sterile
포장
pack of 10 ea (double bagged)
제조업체/상표
Bel-Art H36940-1010
길이 × 너비
17.8 cm (7 in.) × 0.8125 in. (20 mm)
공칭 용량
1.25 mL , ¼tsp
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
특징 및 장점
  • Ten individually packaged spoons are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10-6
  • Lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)