Key Documents
OH83033969
Ohaus® pH Electrode
STPURE
Select a Size
1 EACH
₩264,712
₩264,712
Estimated to ship onMay 07, 2025Details
Select a Size
Change View
1 EACH
₩264,712
About This Item
UNSPSC Code:
41115604
₩264,712
Estimated to ship onMay 07, 2025Details
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 83033969
pH measuring range
0.00-14 pH unit
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)