Key Documents
OH30240974
Ohaus® pH Electrode
ST270
Select a Size
1 EACH
₩563,731
₩563,731
Estimated to ship onApril 02, 2025Details
Select a Size
Change View
1 EACH
₩563,731
About This Item
UNSPSC Code:
41115604
₩563,731
Estimated to ship onApril 02, 2025Details
material
epoxy shaft
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30240974
pH measuring range
0.00-14 pH unit
temperature measuring range
0.0-100 °C
General description
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Features and Benefits
Can be used in conjunction with all Starter bench and portable meters
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)