Skip to Content
Merck
All Photos(1)

Documents

HPA013350

Sigma-Aldrich

Anti-PTPN22 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2

Synonym(s):

Anti-Lyp, Anti-Lyp1, Anti-Lyp2, Anti-PTPN8, Anti-bA402L1.8, Anti-protein tyrosine phosphatase, non-receptor type 22 (lymphoid)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SLLNQESAVLATAPRIDDEIPPPLPVRTPESFIVVEEAGEFSPNVPKSLSSAVKVKIGTSLEWGGTSEPKKFDDSVILRPSKSVKLRSPKSELHQDRSSPPPPLPERTLESFFLADEDCMQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPN22(26191)

Immunogen

protein tyrosine phosphatase, non-receptor type 22 (lymphoid) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PTPN22 (protein tyrosine phosphatase, non-receptor type 22) gene encodes a member of the non-receptor class 4 subfamily belonging to the protein-tyrosine phosphatase family. The encoded protein is a lymphoid-specific intracellular phosphatase that negatively regulates T-cell receptor (TCR) signaling. It dephosphorylates the Src family kinases and certain other key signaling molecules involved in TCR signaling. It functions in the development and activation of lymphocytes, and in innate immune cell–mediated host defense and immunoregulation. The gene is localized to human chromosome 1p13, a region associated with the incidence of type 1 diabetes in certain populations. Mutations in this gene are linked to increased susceptibility to autoimmunity and infection.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70031

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nunzio Bottini et al.
Annual review of immunology, 32, 83-119 (2013-12-25)
Inheritance of a coding variant of the protein tyrosine phosphatase nonreceptor type 22 (PTPN22) gene is associated with increased susceptibility to autoimmunity and infection. Efforts to elucidate the mechanisms by which the PTPN22-C1858T variant modulates disease risk revealed that PTPN22
Xiao Yu et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(50), 19767-19772 (2007-12-07)
The lymphoid-specific tyrosine phosphatase (Lyp) has generated enormous interest because a single-nucleotide polymorphism in the gene (PTPN22) encoding Lyp produces a gain-of-function mutant phosphatase that is associated with several autoimmune diseases, including type I diabetes, rheumatoid arthritis, Graves disease, and
Nunzio Bottini et al.
Seminars in immunology, 18(4), 207-213 (2006-05-16)
We recently discovered that a single-nucleotide polymorphism (SNP) in the lymphoid tyrosine phosphatase (LYP), encoded by the PTPN22 gene on chromosome 1p13, correlates strongly with the incidence of type 1 diabetes (T1D) in two independent populations. This findings has now
Jiansheng Wu et al.
The Journal of biological chemistry, 281(16), 11002-11010 (2006-02-08)
Stimulation of mature T cells activates a downstream signaling cascade involving temporally and spatially regulated phosphorylation and dephosphorylation events mediated by protein-tyrosine kinases and phosphatases, respectively. PTPN22 (Lyp), a non-receptor protein-tyrosine phosphatase, is expressed exclusively in cells of hematopoietic origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service