HPLCによるクマフリルエナンチオマーの分析、 CHIROBIOTIC® V
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 20 mM ammonium nitrate, pH 5.5; [B] THF; (90:10, A:B)
flow rate
1.2 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
2 mg/mL in methanol