HPLCによるジペロドンエナンチオマーの分析、Astec® Cellulose DMP (no additives)
![HPLCによるジペロドンエナンチオマーの分析、Astec® Cellulose DMP (no additives) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/416/008/88eff3c7-207f-4439-8b49-a531ca0640f3/800/88eff3c7-207f-4439-8b49-a531ca0640f3.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] acetonitrile; (100, A)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in mobile phase