Merck
HPLC Analysis of Horse Serum Proteins in Acetonitrile as a Function of Ammonium Formate Concentration
HPLC Analysis of Horse Serum Proteins in Acetonitrile as a Function of Ammonium Formate Concentration application for HPLC
材料
ガードカラム
製品番号
詳細
価格
SRT® SEC-150 Gel Filtration Guard Column, 5 μm
L × I.D. 5 cm × 4.6 mm, 5 μm particle size, stainless steel
分析カラム
製品番号
詳細
価格
SRT® SEC-150 Gel Filtration Column, 5 μm
L × I.D. 30 cm × 4.6 mm, 5 μm particle size, stainless steel
CONDITIONS
column
SRT SEC-150, 30 cm x 4.6 mm I.D., 5 μm particles, 150 Å (Z777043)
mobile phase
100–200 mM CH3COONH4/CH3CN, pH 6.3
flow rate
0.25 mL/min
detector
SofTA ELSD
sample
Molecular weight marker proteins, horse serum
詳細
アナリシスノート
Although horse serum proteins can be analyzed under standard SEC conditions, the resolution is poor. However, very high efficiency separations can be achieved when using pure acetonitrile to which ammonium formate has been added. About 200 mM ammonium formate is required for all proteins to elute within the pore volume of SRT SEC-150 column. The position of the components can be modulated by decreasing the ammonium formate concentration.
Ref.: V. McKay; X. Huang, LC-GC North America;Sep2006 Supplement, Vol. 24, p25, 2D Chromatography of Proteins: Horse Serum Analysis
その他情報
Courtesy of Miyako Kawakatsu, M&S Instruments Inc.