24417
cone tip stainless steel needle (point style 1)
26 ga
10R-HP-0.47color (dark orange)
needle type R (removable needle)syringe type Standard syringe, removable needle
pkg of 1 ea
SGE 002805
42 mm
10 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
最新バージョンのいずれかを選択してください:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 資料 section.