Merck
Z742700
BenchMixer V2 Vortexer with reversible top
AC/DC input 115 V AC, US 2-pin plug
別名:
vortex mixer, vortexer lab, vortexer mixer
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥64,950
¥64,950
出荷予定日2025年4月16日
サイズを選択してください
表示を変更する
1 EA
¥64,950
About This Item
UNSPSCコード:
41103800
NACRES:
NB.73
¥64,950
出荷予定日2025年4月16日
包装
pack of 1 ea
メーカー/製品名
Benchmark Scientific BV1003
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The BenchMixer V2 utilizes a smooth and powerful mixing technology similar to the original BenchMixer, with the advantage of a uniquely designed reversible rubber head that can be attached to the mixer in two orientations.
For the vortexing of single tubes, the cupped surface would generally be preferred. However, for applications requiring a flat/dimpled surface for mixing uniquely shaped vessels (or a handful of multiple test-tubes), the head can easily be flipped over and reattached in reverse orientation.
Like the original BenchMixer, the drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. With a maximum speed up to 3,200 rpm, the mixer provides an instant vortex of even the largest sample sizes, including full 50ml tubes.
特徴および利点
Reversible top platform, cup or flat-dimpled surface
Powerful motor for INSTANT vortexing
Continuous or "touch" operation
Q-Drive dynamic balancing system
Variable speed from 200 to 3200 rpm
Wide variety of accessories, for tubes up to 50ml
法的情報
Benchmixer is a trademark of Benchmark Scientific, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)