サイズを選択してください
960 EA
¥17,320
¥17,320
出荷予定日2025年4月12日
すべての画像(2)
サイズを選択してください
表示を変更する
960 EA
¥17,320
About This Item
UNSPSCコード:
41121600
NACRES:
NB.01
最大容量:
1.2 mL
容量範囲:
50-1200 μL
無菌性:
non-sterile
特徴:
DNase free
RNase free
barrier: no
endotoxin free
RNase free
barrier: no
endotoxin free
包装:
pack of 96 ea (Racked)
メーカー/製品名:
Sartorius 791200
¥17,320
出荷予定日2025年4月12日
おすすめの製品
物質
colorless
無菌性
non-sterile
特徴
DNase free
RNase free
barrier: no
endotoxin free
包装
pack of 96 ea (Racked)
メーカー/製品名
Sartorius 791200
最大容量
1.2 mL
容量範囲
50-1200 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus Nxt, Picus, Tacta, Mline and Proline Plus pipettes.
Optifit Tips are available in various packaging types for all your needs:
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Optifit Tips are available in various packaging types for all your needs:
- Single Tray racks – purity certified tip racks
- Refill Packs - purity certified and ecological refill packages
- Refill Towers - space-saving and ecological refill packages
- Flexibulk boxes - space-saving, ecological and economical bulk boxes
- Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
その他情報
Browse all pipette tips
最新バージョンのいずれかを選択してください:
アクティブなフィルタ