Merck
Z709859
Sartorius pipette tips
volume range 0.1-10 μL, standard, racked, non-sterile
ログイン組織・契約価格を表示する
サイズを選択してください
960 EA
¥17,320
¥17,320
Check Cart for Availability
サイズを選択してください
表示を変更する
960 EA
¥17,320
About This Item
UNSPSCコード:
41121600
NACRES:
NB.01
¥17,320
Check Cart for Availability
物質
colorless
無菌性
non-sterile
特徴
DNase free
RNase free
barrier: no
endotoxin free
包装
pack of 96 ea (Racked)
メーカー/製品名
Sartorius 790010
最大容量
10 μL
容量範囲
0.1-10 μL
容量
0.1—10 μL
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
その他情報
Browse all pipette tips
最新バージョンのいずれかを選択してください:
試験成績書(COA)