Merck
Z379417
Sigma-Aldrich
Hanna Piccolo® pH meter
Piccolo 2
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41115603
詳細
temperature compensation 0 - 70 °C (automatic with integral temperature sensor)
特徴
calibration manual dual point, ±1 pH
メーカー/製品名
HI98112
pH測定範囲
0-14, accuracy: 0.01, resolution: 0.01
電極長
160 mm
重量
400 gincluding calibration screwdriver, ABS carrying case, and batteries
70 g
バッテリー電源
3 × 1.4 V button cell, 100 hr life (continuous operation)
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Hand-held stick-style pH/temperature meters with rugged design for field use. "Amplified electrode" delivers strong signal to the meter, eliminating the vulnerability to noise, humidity, and dirt that is characteristic of conventional meters.
法的情報
Piccolo is a registered trademark of Hanna Instruments
最新バージョンのいずれかを選択してください:
試験成績書(COA)