Merck
Z376213
Sigma-Aldrich
PCR Workstation
W × D × H 48 in. × 24 in. × 24 in., AC/DC input 120 V AC
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41100000
NACRES:
NB.22
テクニック
PCR: suitable
W × D × H
48 in. × 24 in. × 24 in.
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
A self-contained work area that will help protect your PCR runs against contamination. Dual UV bulbs irradiate the work area prior to use, reducing the possibility that contaminating DNA will be amplified. Containment features reduce the chance of airborne contamination.
The non-ventilated hood has acrylic windows for a clear view, an acid-resistant black formica work surface, a laminated safety glass fascia, and double access doors. The germicidal lamps are controlled by a 24-hr timer. The work light is provided by dual fluorescent lamps, and there is a ceiling-mounted double AC outlet. Replacement part sold separately.
法的情報
PCR Workstation is a trademark of C.B.S. Scientific Co.
最新バージョンのいずれかを選択してください:
試験成績書(COA)