Merck
VIV105007STU3
SynVivo SynTumour 3D cancer model chip, microvascular network configuration, 2 um pillar
new
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
60104506
NACRES:
NA.84
現在、価格および在庫状況を閲覧できません。
包装
case of 3 ea
メーカー/製品名
SynVivo 105007-STU3
保管温度
room temp
詳細
SynTumor cancer model chip- Microvascular design Barrier type: Pillars, Distance between pillars: 3 μm, Pillar Diameter: 20 μm, Barrier height: 2 μm, Travel/barrier width: 50 μm, Device depth: 100 μm. SynTumor is a 3D tissue model for real-time visualization and quantitative assessment of cell-cell and cell-drug interactions in a physiologically and morphologically realistic tumor microenvironment.
関連事項
LInk to Publications application text
最新バージョンのいずれかを選択してください:
試験成績書(COA)