表示を変更する
| サイズ/SKU | 在庫状況 | 単価 |
|---|---|---|
500 ea | 在庫from市川市大町368 | ¥24,100 |
この商品について
NACRES:
NB.22
UNSPSC Code:
41122103
¥24,100
在庫詳細
sterility
sterile
feature
individually wrapped
packaging
pk of 500
manufacturer/tradename
Heathrow Scientific
technique(s)
cell culture: suitable
size × width × thickness
172 mm × 37 mm × 4 mm
Features and Benefits
- Smooth spreading is aided by a domed headpiece.
- Agar tears are reduced with a flexible rippled handle and domed spreading bar.
- Disposable
- γ sterilized
Legal Information
Delta is a trademark of Heathrow Scientific LLC
1 of 1
当該品目 | ||
|---|---|---|
| description individually wrapped, sterile, pk of 500 | description L-Shape, sterile, pack of 500 ea (Packed as 50 x bags of 10ea) | description AC/DC input 230 V AC, UK plug |
| feature individually wrapped | feature - | feature - |
| sterility sterile | sterility sterile | sterility - |
| packaging pk of 500 | packaging pack of 500 ea (Packed as 50 x bags of 10ea) | packaging - |
| manufacturer/tradename Heathrow Scientific | manufacturer/tradename Heathrow Scientific HS8171B | manufacturer/tradename Benchmark Scientific MR9600-E-UK |
| size × width × thickness 172 mm × 37 mm × 4 mm | size × width × thickness - | size × width × thickness - |