表示を変更する
| サイズ/SKU | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥15,700 |
この商品について
NACRES:
NB.17
UNSPSC Code:
41121800
Manufacturer/tradename:
Heathrow Scientific HS23403
Feature:
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
Material:
acrylic , clear (bin)
Packaging:
pack of 1 ea
¥15,700
カートの在庫状況を確認する
material
acrylic , clear (bin)
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23403
L × W × H
217 mm × 170 mm × 217 mm
General description
The Workstation Dispensing Bin′s crystal-clear bins let you see your small items inventory.
Features and Benefits
- Three Compartments
- See inventory levels of pipette tips, tubes, fittings, finger cots, and other bulk small items at a glance
- Flip-top covers on generous. openings make filling and dispensing easy and keep products dust-free
- Includes non-skid rubber feet
1 of 1
当該品目 | |||
|---|---|---|---|
| description clear (bin) | description clear (bin) | description blue acrylic | description capacity 100 mL |
| feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature - | feature - |
| manufacturer/tradename Heathrow Scientific HS23403 | manufacturer/tradename Heathrow Scientific HS23402 | manufacturer/tradename Heathrow Scientific | manufacturer/tradename Ace Glass 723930 |
| material clear (bin), acrylic | material clear (bin), acrylic | material blue acrylic | material - |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging - | packaging - |
| L × W × H 217 mm × 170 mm × 217 mm | L × W × H 150 mm × 177 mm × 368 mm | L × W × H - | L × W × H - |