Merck
GBL665273
Grace Bio-Labs CultureWellシリコンアイソレータシート材料
thickness 1.5 mm, (Adhesive on one side)
別名:
silicone sheet with single side adhesive
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41161502
NACRES:
NB.22
物質
clear silicon
無菌性
non-sterile
特徴
(Adhesive on one side)
包装
pack of 5 ea
メーカー/製品名
Grace Bio-Labs 665273
L × W
180 mm × 130 mm
厚さ
1.5 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
CultureWell clear silicone sheet material allows researchers to create their own removable hydrophobic barriers to isolate specimens. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides.
法的情報
Culturewell is a trademark of Grace Bio-Labs, Inc.