表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|
この商品について
UNSPSC Code:
41122405
NACRES:
NB.35
eCl@ss:
32035504
現在、価格および在庫状況を閲覧できません。
material
stainless steel
packaging
pack of 1 ea
L
6 in.
General description
with serrated tips
1 of 1
当該品目 | |||
|---|---|---|---|
| material stainless steel | material stainless steel | material stainless steel | material stainless steel |
| L 6 in. | L 4 3/4 in. | L 12 in. | L 10 in. |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Dressing tissue forceps