Merck
BAF133550001
Fluorescent Ruler, Metric/English Scale
pink phosphorescent ink visible under UV
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥4,650
¥4,650
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EA
¥4,650
About This Item
UNSPSCコード:
41106000
NACRES:
NB.22
¥4,650
Check Cart for Availability
メーカー/製品名
Bel-Art F13355-0001
測定範囲
0-15 cm
詳細
This plastic metric/English scale is printed with a highly visible, glow-in-the-dark pink phosphorescent ink. Under UV light the ruler graduations glow visibly and help to calculate nucleic acid mobility. The ruler is marked 0 to 15cm in millimeter increment, along one edge and 0" to 6" in 1/16" increments on the other edge.
High visibility, glow-in-the-dark scale
  • Ideal for viewing fluorescent images and fluorescent photography
最新バージョンのいずれかを選択してください:
試験成績書(COA)