Merck
MA0HC23CL3
Millipore
ミリスタックプラス(Millistak+)® デプスフィルター μ POD形式
(A0HC media series), cellulose membrane (and inorganic filter aid), effective filtration area 23 m2
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
23151806
物質
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
品質水準
滅菌適合性
autoclavable compatible
有効ろ過面積
23 m2
フィッティング
seal (flat seal)
カートリッジコード
(A0HC media series)
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Millistak+® A0HC media series Depth Filters is offered in a scalable, disposable format. Accommodating applications from lab to pilot to process scale, the μPod format offers greater flexibility because of its unique modular design.
Millistak+® μPod system is ideal for a wide variety of Primary and Secondary Clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
The Millistak+® Pod Filters are available in three distinct series of media grades in order to meet your specific application needs. The Millistak+® A0HC media series delivers optimal performance through a gradient density matrix as well as positive surface charge properties.
法的情報
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany