Syndecan-4 (Amphiglycan; Ryudocan core protein) is one of four members of the syndecan proteoglycan family. The syndecan proteins bind heparan sulfate and mediate the signaling of growth factors, such as fibroblast growth factor, that recognize and bind heparan chains. Unlike other members of this family, syndecan-4 is ubiquitously expressed, and is targeted to focal adhesions, possibly by a PKC-mediated pathway. Previous studies have reported an increase in focal adhesions in cells overexpressing syndecan-4. This protein also plays an important role in wound healing, by modulating the signaling of Rho-GTPases Rac1, RhoG, and RhoA.
特異性
This antibody recognizes the cytoplasmic domain of Syndecan-4
免疫原
Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Syndecan-4.
アプリケーション
Research Category 細胞骨格
Research Sub Category 細胞骨格シグナル伝達
Immunohistochemistry Analysis: A 1:100-1,000 dilution from a representative lot detected Syndecan-4 in human lung carcinoma and in distal tubules of normal human kidney tissue.
This Anti-Syndecan-4 Antibody is validated for use in Western Blotting, IHC(P) for the detection of Syndecan-4.
品質
Evaluated by Western Blot in HEP2 cell lysate.
Western Blot Analysis: 1 µg/mL of this antibody detected Syndecan-4 in 10 µg of HEP2 cell lysate.
ターゲットの説明
~80/32 kDa observed
物理的形状
Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
保管および安定性
Stable for 1 year at 2-8°C from date of receipt.
アナリシスノート
Control HEP2 cell lysate
その他情報
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
免責事項
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Cardiovascular diseases are a group of disorders caused by the presence of a combination of risk factors, such as tobacco use, unhealthy diet and obesity, physical inactivity, etc., which cause the modification of the composition of the vessel's matrix and
Journal of cellular biochemistry, 120(8), 13826-13840 (2019-04-05)
Anoikis is a form of programmed cell death induced by loss of contact from neighboring cells or from their extracellular matrix (ECM). Many tumorigenic cells are anoikis resistant, facilitating cancer progression and metastasis. Trastuzumab is a monoclonal antibody used for
Matrix biology : journal of the International Society for Matrix Biology, 70, 102-122 (2018-04-02)
Intervertebral disc degeneration and associated low back and neck pain is a ubiquitous health condition that affects millions of people world-wide, and causes high incidence of disability and enormous medical/societal costs. However, lack of appropriate small animal models with spontaneous
Aldosterone contributes to end-organ damage in heart failure and chronic kidney disease. Mineralocorticoid-receptor inhibitors limit activation of the receptor by aldosterone and slow disease progression, but side effects, including hyperkalemia, limit their clinical use. Damage to the endothelial glycocalyx (a