Z115517
1 of 10
Supelco
1.09535
pH-indicator strips pH 0 - 14 Universal indicator
Sigma-Aldrich
Z115509
Scienceware® protective lab-labeling system tape
Z115487
Z334235
BRAND® test tube racks, PP
1.04699
Immersion oil
Millipore
1.04140
Singlepath® Salmonella
1.06586
リン酸ナトリウム 二塩基性
L8144
Sigma label tape
HS159890B
Storage rack for 50 and 100 place microscope slide boxes
1.00609
Nitrite Cell Test
paper white
pkg of 10 (100 labels per pad)
Bel-Art F13455-0005
1/2 in. × 1 1/2 in.
white
類似した製品をお探しですか? 訪問 製品比較ガイド
最新バージョンのいずれかを選択してください:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 資料 section.