Merck
S941530
5'-CHLORO-4'-METHOXY-2'-NITROACETANILIDE
AldrichCPR
ログイン組織・契約価格を表示する
サイズを選択してください
50MG
¥24,200
¥24,200
出荷予定日2025年5月21日
サイズを選択してください
表示を変更する
50MG
¥24,200
About This Item
化学式:
C9H9ClN2O4
CAS番号:
分子量:
244.636
MDL番号:
¥24,200
出荷予定日2025年5月21日
その他情報
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
最新バージョンのいずれかを選択してください:
試験成績書(COA)