Merck
R425249
(4-OXO-3-PHENYL-2-PHENYLIMINO-THIAZOLIDIN-5-YL)-ACETIC ACID
AldrichCPR
ログイン組織・契約価格を表示する
サイズを選択してください
R425249-100MG
¥21,500
¥21,500
出荷予定日2025年4月24日
サイズを選択してください
表示を変更する
R425249-100MG
¥21,500
About This Item
化学式:
C17H14N2O3S
CAS番号:
分子量:
326.377
MDL番号:
¥21,500
出荷予定日2025年4月24日
その他情報
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
最新バージョンのいずれかを選択してください:
試験成績書(COA)