おすすめの製品
AC/DC入力
(US/UK/Australia /Scandanavia AC interchangable input plugs)
特徴
capacity 16 vial(s)
パラメーター
435-445 nm spectral range
関連するカテゴリー
詳細
特徴および利点
- 【キット内容】
- アルミニウム製サンプルホルダー(直径11 cm):4 mLバイアル16個をセット可能
- 青色LEDライトリング(波長:435-445 nm)、防水仕様(IP68規格)
- 【推奨品(別売)】
- バイアル:キャップ付バイアル(27138)、またはセプタムキャップ付バイアル(27209-U)
- 攪拌子(10 x 3 mm):Z118850
- 紫色LEDライトリング(波長:400-410 nm):ALDRP2
- 国内販売品に電源アダプターはついておりません。別途ご用意ください。(規格は下記Other Notes参照)
法的情報
関連製品
試験成績書(COA)
この製品を見ている人はこちらもチェック
資料
Photocatalysis in organic synthesis has grown with innovative methods using visible light, contributing to new research.
近年、光レドックス触媒と可視光を利用した可視光応答型反応の研究が進み、これまで実現していなかった新規合成法が次々と報告されています。
This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.
This article details the latest progress in synthesizing alkylsulfonyl fluorides through different approaches that utilize photoredox catalysis, electrocatalysis, transition-metal catalysis, and organocatalysis. The alkylsulfonyl fluorides thus prepared could be utilized further in sulfur(VI) fluoride exchange (SuFEx) click reactions.
関連コンテンツ
One major theme of research in the Yoon lab is the development of unique chemical transformations enabled by visible light photochemistry. A variety of transition metal photocatalysts responsive to visible light are known, and the Yoon lab has shown that their well understood photochemistry can be coupled to cycloadditions, arene oxidations, amine functionalizations, and thiol-ene reactions, among others.