Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

233501

Sigma-Aldrich

4-Hexylaniline

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H4NH2
CAS Number:
Molecular Weight:
177.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.525 (lit.)

bp

279-285 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCCCCCc1ccc(N)cc1

InChI

1S/C12H19N/c1-2-3-4-5-6-11-7-9-12(13)10-8-11/h7-10H,2-6,13H2,1H3

InChI key

OVEMTTZEBOCJDV-UHFFFAOYSA-N

General description

4-Hexylaniline reacts with isatin (1H-indole-2,3-dione) to form Schiff base ligand (HL). Mesomorphic properties of imine compounds derived from 4-hexylaniline has been investigated using polarised light optical microscopy and differential scanning calorimetry.

Application

4-Hexylaniline has been used in the preparation of stable ruthenium nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

233501-50G:
233501-25G:
233501-VAR:
233501-5G:
233501-10G:
233501-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solid-state electronic conductivity of ruthenium nanoparticles passivated by metal-carbon covalent bonds.
Ghosh D and Chen S.
Chemical Physics Letters, 465(1), 115-119 (2008)
Novel Isatin-Schiff Base Cu (H) and Ni (H) Complexes. X-ray Crystal Structure of Bis [3-(4-hexylphenylimino)-1H-indol-2 (3H)-one]-dichlorocopper (H) Complex.
Ercag A, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 17(2) (2006)
Liquid crystals based on semiperfluorinated imines and salicylaldimato metal complexes. A comparative study of alkyl, alkoxy and polyether substituents.
Bilgin-Eran B, et al.
Journal of Materials Chemistry, 17(22), 2319-2328 (2007)
Kyle A Brown et al.
Nature methods, 16(5), 417-420 (2019-04-17)
We report the identification of a photocleavable anionic surfactant, 4-hexylphenylazosulfonate (Azo), which can be rapidly degraded by ultraviolet irradiation, for top-down proteomics. Azo can effectively solubilize proteins with performance comparable to that of sodium dodecyl sulfate (SDS) and is compatible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service