- A photocleavable surfactant for top-down proteomics.
A photocleavable surfactant for top-down proteomics.
Nature methods (2019-04-17)
Kyle A Brown, Bifan Chen, Tania M Guardado-Alvarez, Ziqing Lin, Leekyoung Hwang, Serife Ayaz-Guner, Song Jin, Ying Ge
PMID30988469
ABSTRACT
We report the identification of a photocleavable anionic surfactant, 4-hexylphenylazosulfonate (Azo), which can be rapidly degraded by ultraviolet irradiation, for top-down proteomics. Azo can effectively solubilize proteins with performance comparable to that of sodium dodecyl sulfate (SDS) and is compatible with mass spectrometry. Azo-aided top-down proteomics enables the solubilization of membrane proteins for comprehensive characterization of post-translational modifications. Moreover, Azo is simple to synthesize and can be used as a general SDS replacement in SDS-polyacrylamide gel electrophoresis.