Documenti
Z227285
Supelco® PEEK Fitting
Two-piece with ferrule, fingertight/wrenchtight
About This Item
Codice UNSPSC:
41115700
NACRES:
SB.52
Confezionamento
pkg of 5 ea
Descrizione generale
Biocompatible fittings for all 1/16 in. tubing connections.
Caratteristiche e vantaggi
Two-Piece Fingertight/Wrenchtight Fittings: Hold to 6000 psi (420kg/cm2) when finger-tightened or 10,000 psi (700kg/cm2)when wrenchtightened. Double-tapered ferrules included.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)