Documenti fondamentali
22116
Fine Metering Valve
new
brass, 1/8 in. straight arms
About This Item
Codice UNSPSC:
41115717
eCl@ss:
32035302
Materiali
brass
Descrizione
fine adjustment
Raccordi
1/8 in. straight arms
Descrizione generale
Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.
Applicazioni
Fine metering valve may be used on the exit of a flow system, which is basically used for the conductance measurements of decyl-, dodecyl-, tetradecyl- and hexadecyltrimethylammonium bromide in water at 25, 60, 95, 130, and 160°C, primarily done in order to determine their critical micelle concentrations (CMCs).[1]
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Paulami Bose et al.
Nanoscale, 12(43), 22116-22128 (2020-10-30)
Reactions between atomically precise noble metal nanoclusters (NCs) have been studied widely in the recent past, but such processes between NCs and plasmonic nanoparticles (NPs) have not been explored earlier. For the first time, we demonstrate spontaneous reactions between an
Critical micelle concentrations for alkyltrimethylammonium bromides in water from 25 to 160 C.
Fennell ED
Journal of Solution Chemistry, 13 (2), 87-101 (1984)