Documenti fondamentali
22063
Compact Adapter
About This Item
Codice UNSPSC:
41115717
Descrizione
compact reducer
Confezionamento
pkg of 1 ea
Descrizione generale
Use as a 1/4 in. to 1/16 in. Swagelok reducer,
or as a 1/4 in. to 1/16 in. capillary I.D. reducing ferrule.
Also fits Varian capillary injection ports. Stainless steel body. Includes 1/4 in. M-2A ferrule. Purchase appropriate capillary ferrules separately.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)