Documenti fondamentali
Z740431
HistoTainer II prefilled specimen containers
M961, volume 40 mL
Scegli un formato
96 EA
119,00 €
119,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
96 EA
119,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
119,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polyethylene cap
polypropylene container
Confezionamento
pack of 96 ea
Produttore/marchio commerciale
Simport M961-40FMA
Volume
40 mL
Descrizione generale
NON Tamper Evident Prefilled Specimen Containers, 50% filled with 10% Neutral Buffered Formalin
Especially designed for collection, transport and storage of histology specimens, Simport offers shatter resistant polypropylene containers, eliminating most problems of leakage and evaporation. Containers are manufactured from virgin, translucent polypropylene. The white lids are ribbed for easy opening when hands are wet or gloved while the jars are stackable for easy, safe storage. Jars are translucent and specimens can be viewed without having to open the lid. Closures are manufactured from virgin polyethylene with a unique integrated leak-resistant seal.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All materials used in manufacturing are free from latex. Containers are
95 kPa compliant. Available in many sizes from 20 to 120 ml. Packaging is offered in trays of 24, cases of 96.
The HistoTainer II is half filled with a choice of fixatives. 10% Neutral Buffered Formalin penetrates quickly, but fixes slowly. The Formalin is enhanced by a buffering capacity optimizing histological results by light microscopy and immunohistochemistry. Bouin, Hollande and Zinc Formalin fixatives are also available.
Note legali
HistoTainer is a trademark of Simport
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)