TMO3110-9500
Nalgene® high-speed centrifuge tubes
round bottom natural polypropylene copolymer, volume 50 mL, without cap, non-sterile, case of 100 ea
Sinonimo/i:
Nalgene tubes, round bottom tubes
About This Item
Codice UNSPSC:
41121703
NACRES:
NB.11
Prodotti consigliati
Materiali
round bottom natural polypropylene copolymer
Sterilità
non-sterile
Caratteristiche
without cap
Confezionamento
case of 100 ea
Produttore/marchio commerciale
Nalgene 3110-9500
Volume
50 mL
Categorie correlate
Descrizione generale
Improve rotor fit and performance using Thermo Scientific Nalgene High-Speed Round-Bottom PPCO Centrifuge Tubes, featuring uniform wall thickness and diameter. Tubes have excellent chemical resistance.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)