Vai a
T2168
Nalgene® centrifuge tubes, Oak Ridge Style 3114
capacity 10 mL, pack of 2 ea
Sinonimo/i:
High-Speed ​​Centrifuge Tubes, Nalgene 3114-0010, sample collection tube, sample receptacle, sample tube
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
A voi/SKUDisponibilitàPrezzo
2 ea
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
97,50 €
Informazioni su questo articolo
NACRES:
NB.11
UNSPSC Code:
41121703
Capacity:
10 mL
Material:
FEP , Tefzel® closure, round bottom tube, screw top cap
Sterility:
non-sterile
Packaging:
pack of 2 ea
Feature:
RCF 10,000 × g
97,50 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
FEP , Tefzel® closure, round bottom tube, screw top cap
sterility
non-sterile
feature
RCF 10,000 × g
packaging
pack of 2 ea
manufacturer/tradename
Nalgene® 3114-0010
capacity
10 mL
General description
Conduct high-speed centrifugation of aggressive chemicals using Nalgene® Oak Ridge High-Speed Centrifuge Tubes made with PTFE® fluoropolymer and Closure made with Tefzel®, specifically suited for chloroform and phenol extractions.
Features and Benefits
  • temperature range -100° to +150°C (-148° to +302°F).
  • Leakproof† closures prevent material loss under ordinary use for most spins <10,000 × g.
  • When equipped with a Nalgene® sealing cap assemblies30mL and 50mL tubes can be spun up to 50,000 x g.
  • Must be filled to at least 100% total capacity for proper performance.
  • Autoclavable/Biohazard/Leakproof† when used with supplied Nalgene® closure /PTFE FEP.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
T2418T2293T3293
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
Nalgene® 3114-0010
manufacturer/tradename
Nalgene® 3114-0050
manufacturer/tradename
Nalgene® 3114-0030
manufacturer/tradename
Nalgene® 3138-0030
material
FEP , round bottom tube, Tefzel® closure, screw top cap
material
FEP , Tefzel® closure, round bottom tube, screw top cap
material
FEP , Tefzel® closure, round bottom , screw top cap
material
cap (PP sealing cap), round bottom tube, transparent polycarbonate
capacity
10 mL
capacity
50 mL
capacity
30 mL
capacity
-
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 10 ea
feature
RCF 10,000 × g
feature
RCF 50,000 × g with sealing cap assembly
feature
RCF 50,000 × g with sealing cap assembly
feature
RCF 50,000 × g
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)