Scegli un formato
| A voi/SKU | Disponibilità | Prezzo |
|---|---|---|
1 ea | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 111,00 € |
Informazioni su questo articolo
111,00 €
material
acrylic , clear (bin)
feature
holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS23404
L × W × H
287 mm × 170 mm × 217 mm
General description
Features and Benefits
- Four Compartments
- See inventory levels of pipette tips, tubes, fittings, finger cots, and other bulk small items at a glance
- Flip-top covers on generous. openings make filling and dispensing easy and keep products dust-free
- Includes non-skid rubber feet
1 of 1
Questo articolo | |||
|---|---|---|---|
| description clear (bin) | description clear (bin) | description clear (bin) | description blue |
| feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Pipette tips, tubes, finger cots, fittings, and other small bulk items, accessories | feature holds Small laboratory consumables |
| manufacturer/tradename Heathrow Scientific HS23404 | manufacturer/tradename Heathrow Scientific HS23403 | manufacturer/tradename Heathrow Scientific HS23402 | manufacturer/tradename Heathrow Scientific HS23412 |
| material acrylic , clear (bin) | material acrylic , clear (bin) | material acrylic , clear (bin) | material acrylic , blue |
| L × W × H 287 mm × 170 mm × 217 mm | L × W × H 217 mm × 170 mm × 217 mm | L × W × H 150 mm × 177 mm × 368 mm | L × W × H 155 mm × 155 mm × 170 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?