Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
material
square white polypropylene
feature
to hold 16 x 1,5/2.0 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Heathrow Scientific HS2132A
General description
Features and Benefits
- Square Shape Rack
- 1.5/2.0 mL Tubes, 16 Wells (10.9 mm well dia.)
- Use on the bench, with water baths or ice containers
- Keeps microtubes afloat in a water bath or can be pushed into crushed ice — perfect for controlling sample temperature
- Attach legs so rack stands upright on bench top
- Detach legs so samples can be quickly removed by pushing down onto any surface
- Not recommended for autoclaving
1 of 1
Questo articolo | |||
|---|---|---|---|
| description to hold, 16 x 1,5/2.0 mL tubes, square white polypropylene | description to hold, 20 x 1,5/2.0 mL tubes, round white polypropylene | description rectangular shaped, holds 24 tubes, Holds 24 x 1.5-2 mL tubes | description parallelogram shaped, holds 8 tubes, Holds 8 x 15 mL tubes |
| feature to hold 16 x 1,5/2.0 mL tubes | feature to hold 20 x 1,5/2.0 mL tubes | feature Holds 24 x 1.5-2 mL tubes | feature Holds 8 x 15 mL tubes |
| manufacturer/tradename Heathrow Scientific HS2132A | manufacturer/tradename Heathrow Scientific HS2134A | manufacturer/tradename Heathrow Scientific HS2165C | manufacturer/tradename Heathrow Scientific HS2165B |
| material square white polypropylene | material round white polypropylene | material yellow , foamed PE | material foamed PE , blue |
| packaging pack of 4 ea | packaging pack of 4 ea | packaging - | packaging - |
Still not finding the right product?
Explore all of our products under Floating Microtube Rack
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?