Vai a
HS1422
Shallow weigh boat
Medium, square white polystyrene, pk of 500 ea
Sinonimo/i:
weigh boat, weigh dish, weighing dish, weighing pan
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
500 ea
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
58,20 €
Informazioni su questo articolo
NACRES:
NB.35
UNSPSC Code:
41121812
eCl@ss:
32040608
Material:
square white polystyrene
Packaging:
pk of 500 ea
Manufacturer/tradename:
Heathrow Scientific HS1422
58,20 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
square white polystyrene
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1422
L × H
90 mm (90 mm) × 7 mm
General description
Our flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
  • Shallow boat with a wide, flat bottom to resist tipping
  • Ideal for solid product weighing
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • Temperature range -10 to 70°C (14 to 158°F)
  • Can be used with microwave moisture analyzers
  • All are FDA/food-grade compliant
  • Pack of 500
  • Disposable
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
HS1423AHS1420AHS1420BB
Shallow weigh boat Medium, square white polystyrene, pk of 500 ea
HS1422
Shallow weigh boat
Weigh Boat Small, square black polystyrene, pk of 500 ea
HS1423A
Weigh Boat
Weigh Boat size small, square white polystyrene
HS1420A
Weigh Boat
Weigh Boat Medium, square white polystyrene, Anti-Static, pack of 500 ea
HS1420BB
Weigh Boat
description
Medium, square white polystyrene, pk of 500 ea
description
Small, square black polystyrene, pk of 500 ea
description
size small, square white polystyrene
description
Medium, square white polystyrene, Anti-Static, pack of 500 ea
manufacturer/tradename
Heathrow Scientific HS1422
manufacturer/tradename
Heathrow Scientific HS1423A
manufacturer/tradename
Heathrow Scientific HS1420A
manufacturer/tradename
Heathrow Scientific HS1420BB
material
square white polystyrene
material
square black polystyrene
material
square white polystyrene
material
square white polystyrene
packaging
pk of 500 ea
packaging
pk of 500 ea
packaging
pack of 500 ea
packaging
pack of 500 ea
L × H
90 mm (90 mm) × 7 mm
L × H
-
L × H
-
L × H
-
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)