Documenti fondamentali
GN664160
Greiner CELLSTAR® dish
100 x 20 mm, 58 cm2, TC treated, sterile, w/ vents
Sinonimo/i:
cell culture dish, cell culture plate, cell culture plates, Greiner CELLSTAR® cell culture dishes
About This Item
Codice UNSPSC:
41121812
Confezionamento
pack of 360 ea
Produttore/marchio commerciale
Greiner 664160
Disponibilità
available only in China
Descrizione generale
The CELLSTAR®culture dish combines the convenience of astandard size 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of invitro cultivated cultures. Made from high-grade polystyrene combined with an integrated glass bottom. The innovative design of the dish provides a single-plane, flat bottom with a consistent working distance and maximal planarity. Moreover, the dish bottom configuration facilitates optimal thermal conductivity andavoids thermal variations in heated platforms used for live cell imaging.
Caratteristiche e vantaggi
  • Greiner CELLSTAR™ dishes are manufactured from highly transparent crystal clear polystyrene for optimal microscopic examinations.
  • Surface treated for optimal cell attachment and maximum cell proliferation.
  • Vented - to facilitate optimal gas exchange.
  • Dishes are stackable and provide a convenient space-saving option.
  • Free from detectable RNA, DNA, endotoxins, and human DNA.
  • Pyrogen-free and non-cytotoxic.
Note legali
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)