Documenti
GN163160
Greiner culture tubes
volume 12 mL, 16 x 100 mm, TC treated, round bottom, sterile, w/ screw cap
Sinonimo/i:
culture tubes, culture vials, plastic culture tubes, Greiner culture tubes
About This Item
Codice UNSPSC:
41121800
Confezionamento
pack of 1000 ea
Produttore/marchio commerciale
Greiner 163160
Disponibilità
available only in China
Descrizione generale
Greiner culture tubes feature a two-position snap vent stopper and graduations with a writing area.