Documenti fondamentali
GBL724550
Grace Bio-Labs HybriSlip hybridization cover
L × W × thickness 45 mm × 50 mm × 0.25 mm, pkg of 250 ea
Sinonimo/i:
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
Scegli un formato
250 EA
683,00 €
683,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
250 EA
683,00 €
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
683,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
colorless
polycarbonate
Confezionamento
pkg of 250 ea
Produttore/marchio commerciale
Grace Bio-Labs 724550
Lungh. × largh. × spess.
45 mm × 50 mm × 0.25 mm
Descrizione generale
HybriSlip ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
  • RNase and DNase free
  • Hydrophobic
  • Disposable
  • Will not chip or break
Note legali
HybriSlip is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)