Documenti fondamentali
GBL622305
Grace Bio-Labs CoverWell perfusion chambers
8 chambers, port diam. 1.5 mm, chamber diam. × thickness 9 mm × 2.5 mm
Sinonimo/i:
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
Scegli un formato
10 EA
110,00 €
110,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
10 EA
110,00 €
About This Item
Codice UNSPSC:
41122600
NACRES:
NB.22
110,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear polycarbonate backing
red silicon wells
Caratteristiche
wells 8
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Grace Bio-Labs 622305
Diam. camera × spessore
9 mm × 2.5 mm
Diametro porta
1.5 mm
Pozzetti
8
Descrizione generale
CoverWell perfusion chambers press-to-seal covers form water-tight, multiwell cell incubation or cytochemistry chambers when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports without disturbing or cross-contaminating specimens. Chambers are available in five chamber configurations allowing up to eight different specimens or analytical tests on one coverglass.
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Note legali
CoverWell is a trademark of Grace Bio-Labs, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)