EPM1289-0300
Test Tube Rack, for Eppendorf® New Brunswick™ Innova® and Eppendorf® Excella® Shakers
holds 42 x 15 mL conical tubes (15-18 mm diam.)
Sinonimo/i:
Quick Change Platform Kit
About This Item
Codice UNSPSC:
41122804
Prodotti consigliati
Caratteristiche
holds 42 x 15 mL conical tubes (15-18 mm diam.)
Produttore/marchio commerciale
Eppendorf® M1289-0300 (Global)
Misura
450x80x129 mm
Categorie correlate
Descrizione generale
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc.
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)