Documenti fondamentali
EP6137928009
Reference Filter Set
for Eppendorf BioSpectrometer® Fluorescence
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Caratteristiche
for Eppendorf BioSpectrometer® Fluorescence
Produttore/marchio commerciale
Eppendorf® 6137928009 (Global)
Descrizione generale
Contains 8 filters (1 blank filter, 3 filters for UV/Vis wavelength systematic error and precision, 3 filters for UV/Vis photometric verification, 1 filter for fluorometric verification)
All features of the Eppendorf BioSpectrometer UV/Vis Reference Filter Set
Additional filter for verifying fluorimetric precision (random error) and linearity
Inspection protocol preprogrammed in device
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf BioSpectrometer is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)