Documenti fondamentali
EP0030627300
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
white polypropylene, V-bottom clear wells, PCR clean, pkg of 240 ea
Scegli un formato
240 EA
1.680,00 €
1.680,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
240 EA
1.680,00 €
About This Item
Codice UNSPSC:
41122107
NACRES:
NB.15
1.680,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
V-bottom clear wells
white polypropylene
Sterilità
non-sterile
Confezionamento
pkg of 240 ea
Produttore/marchio commerciale
Eppendorf® 0030627300
tecniche
PCR: suitable
Volume massimo
-140 μL
Misura
384 wells
Volume d’esercizio
5-120 μL
Tipo di binding
Protein low binding
Descrizione generale
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
The “Eppendorf” Lobind Microplate 384/V-PP is a DNA LoBind plate having 384 wells. It improves the recovery of nucleic acids by reducing their adsorption to the walls of the tube. Special manufacturing techniques along with selected polypropylene batches ensure nearly 100?% recovery of DNA/RNA molecules-without surface coating, thereby eliminating the risk of sample contamination.
Applicazioni
The “Eppendorf” Lobind Microplate 384/V-PP is suitable for:
  • Preparation or storage of DNA and RNA samples
  • Forensic trace analysis
  • Preparing dilution series in quantitative real-time PCR
  • Sample preparation in next-generation sequencing
  • Creation of genomic or oligonucleotide libraries
Caratteristiche e vantaggi
  • The Lobind feature ensures maximum sample recovery thereby improving assay results.
  • The [TM=“Eppendorf”] Lobind Microplate 384/V-PP is devoid of surface coatings to minimize sample interference.
  • Raised well rims and a smooth surface ensure a secure sealing in plates.
  • An optimized [TM=“RecoverMax”] well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • The distinctive [TM=“OptiTrack”] matrix offers accelerated sample identification and reduces pipetting errors.
  • Free of human DNA, DNase, RNase, and PCR inhibitors (PCR clean)
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)